Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKCzeta Rabbit anti-Human, Clone: SN0753, Novus Biologicals™
SDP

Catalog No. NBP267713 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267713 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267713 Supplier Novus Biologicals Supplier No. NBP267713

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

PKC zeta Monoclonal specifically detects PKC zeta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PKCzeta
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN0753
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.13, nPKC-zeta, PKC2PKC-ZETA, protein kinase C zeta type, protein kinase C, zeta
Gene Symbols PRKCZ
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to residues surrounding Thr560 of Human PKC zeta aa 531-580/ 592. (SwissProt: Q05513 Human; SwissProt: Q02956 Mouse; SwissProt: P09217 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Apoptosis, Cancer, Diabetes Research, GPCR, Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5590
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.