Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKHD1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB340241 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340241 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340241 Supplier Novus Biologicals Supplier No. BC11060480APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKHD1 Polyclonal antibody specifically detects PKHD1 in Human, Mouse samples. It is validated for Immunocytochemistry/ Immunofluorescence, ELISA

Specifications

Antigen PKHD1
Applications Immunocytochemistry, ELISA
Classification Polyclonal
Conjugate APC
Dilution Immunocytochemistry/ Immunofluorescence, Peptide ELISA
Formulation PBS
Gene Alias polycystic kidney and hepatic disease 1 (autosomal recessive)
Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt# Q8TCZ9]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5314
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.