Learn More
Description
Specifications
Specifications
| Antigen | PKHD1 |
| Applications | Immunocytochemistry, Immunofluorescence, Peptide ELISA |
| Classification | Polyclonal |
| Conjugate | FITC |
| Dilution | Immunocytochemistry/Immunofluorescence, Peptide ELISA |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | polycystic kidney and hepatic disease 1 (autosomal recessive) |
| Gene Symbols | PKHD1 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt# Q8TCZ9] |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
