Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKHD1 Antibody, Novus Biologicals™
SDP

Catalog No. BC11060480T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BC11060480T 0.025 mL
BC11060480 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BC11060480T Supplier Novus Biologicals Supplier No. BC11060480SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKHD1 Polyclonal specifically detects PKHD1 in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Peptide ELISA.

Specifications

Antigen PKHD1
Applications Immunocytochemistry, Immunofluorescence, Peptide ELISA
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence, Peptide ELISA 1:100-1:2000
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P08F94
Gene Alias polycystic kidney and hepatic disease 1 (autosomal recessive)
Gene Symbols PKHD1
Host Species Rabbit
Immunogen A synthetic peptide derived from the human PKHD1 protein (within residues 3800-3900). [UniProt# Q8TCZ9]
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5314
Test Specificity Human. Immunogen sequence has 86% identity to mouse.
Target Species Human, Mouse, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.