Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PKI-beta Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $499.50
Specifications
Antigen | PKI-beta |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17425520
![]() |
Novus Biologicals
NBP17425520UL |
20 μL |
Each for $158.00
|
|
|||||
NBP174255
![]() |
Novus Biologicals
NBP174255 |
100 μL |
Each for $499.50
|
|
|||||
Description
PKI-beta Polyclonal specifically detects PKI-beta in Mouse, Rat samples. It is validated for Western Blot.Specifications
PKI-beta | |
Polyclonal | |
Rabbit | |
Q8R4R3 | |
5570 | |
Synthetic peptides corresponding to the middle region of Pkib. Immunizing peptide sequence TDVESVISSFASSARAGRRNALPDIQSSLATGGSPDLALKLEALAVKEDA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
cAMP-dependent protein kinase inhibitor 2, cAMP-dependent protein kinase inhibitor beta, FLJ23817, PKI-beta, PRKACN2, protein kinase (cAMP-dependent, catalytic) inhibitor beta | |
PKIB | |
IgG | |
12 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title