Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKLR Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB041847 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB041847 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB041847 Supplier Novus Biologicals Supplier No. NBP297071F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKLR Polyclonal specifically detects PKLR in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PKLR
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate FITC
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PKLR (NP_000289.1; Leu47-Ser574).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chaperone Mediated Autophagy (CMA), Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5313
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.