Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKLR Antibody, Novus Biologicals™
SDP

Catalog No. NBP132314 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132314 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132314 Supplier Novus Biologicals Supplier No. NBP132314
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKLR Polyclonal specifically detects PKLR in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PKLR
Applications Western Blot
Classification Polyclonal
Concentration 0.62mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:10000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias EC 2.7.1.40, PK1PKRL, PKL, PKR, Pyruvate kinase 1, pyruvate kinase isozyme R/L, pyruvate kinase isozymes R/L, pyruvate kinase type L, pyruvate kinase, liver and blood cell, pyruvate kinase, liver and RBC, Red cell/liver pyruvate kinase, RPK, R-type/L-type pyruvate kinase
Gene Symbols PKLR
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Pyruvate Kinase (liver/RBC). The exact sequence is proprietary.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5313
Test Specificity Pyruvate kinase from liver and RBC.
Target Species Human, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.