Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP214833SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214833SS 0.025 mL
NBP214833 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP214833SS Supplier Novus Biologicals Supplier No. NBP214833SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 14 publications

PKM1 Polyclonal specifically detects PKM1 in Bovine, Human, Mouse samples. It is validated for IF/IHC, Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot.

Specifications

Antigen PKM1
Applications Immunoblot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Simple Western, Western Blot
Classification Polyclonal
Concentration 1.17 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Simple Western 1:400, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:400, Immunoblotting reported in scientific literature (PMID 25416385)
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. P14618, P52480
Gene Alias PK1, Pyruvate kinase isozyme M1, Pyruvate kinase isozymes M1, Pyruvate kinase M1
Gene Symbols PKM
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2].
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5315
Test Specificity This antibody is specific for the M1 isoform of PKM.
Target Species Bovine, Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.