Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB004834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB004834 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB004834 Supplier Novus Biologicals Supplier No. NBP214833UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKM1 Polyclonal specifically detects PKM1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting.

Specifications

Antigen PKM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias PK1, Pyruvate kinase isozymes M1, Pyruvate kinase M1
Gene Symbols PKM
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2].
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for the M1 isoform of PKM.
Target Species Human, Mouse, Bovine
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.