Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB337642 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337642 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337642 Supplier Novus Biologicals Supplier No. NBP214833MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKM1 Polyclonal antibody specifically detects PKM1 in Human, Mouse, Bovine samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot

Specifications

Antigen PKM1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting
Formulation 50mM Sodium Borate
Gene Alias PK1, Pyruvate kinase isozymes M1, Pyruvate kinase M1
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human PKM1 protein (between residues 350-450) [UniProt P14618-2].
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5315
Target Species Human, Mouse, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.