Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKM2 Antibody (1B10), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP242686AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242686AH 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242686AH Supplier Novus Biologicals Supplier No. NBP242686AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PKM2 Monoclonal specifically detects PKM2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PKM2
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 1B10
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK
Gene Symbols PKM
Host Species Mouse
Immunogen Recombinant human PKM2 (1-531aa) purified from E.coli.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 5315
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.