Learn More
Description
Ensure accurate, reproducible results in Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Specifications
Specifications
| Antigen | PKM2 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 1B10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:250 |
| Formulation | PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide |
| Gene Accession No. | NP_002645 |
| Gene Alias | CTHBP, Cytosolic thyroid hormone-binding protein, MGC3932, OIP-3, OIP3EC 2.7.1.40, Opa-interacting protein 3, PK, muscle type, PK2, PK3PKM, Pyruvate kinase 2/3, pyruvate kinase isozymes M1/M2, Pyruvate kinase muscle isozyme, pyruvate kinase, muscle, TCB, THBP1p58, Thyroid hormone-binding protein 1, thyroid hormone-binding protein, cytosolic, Tumor M2-PK |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
