Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN1 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB105626 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB105626 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB105626 Supplier Novus Biologicals Supplier No. NBP298761R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKN1 Polyclonal specifically detects PKN1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PKN1
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias DBK, EC 2.7.11, MGC46204, PAK1Serine-threonine protein kinase N, PKNPKN-ALPHA, PRK1PAK-1, PRKCL1protein kinase C-related kinase 1, Protease-activated kinase 1, Protein kinase C-like 1EC 2.7.11.13, Protein kinase C-like PKN, protein kinase N1, Protein kinase PKN-alpha, Protein-kinase C-related kinase 1, serine/threonine protein kinase N, serine/threonine-protein kinase N1, serine-threonine kinase N
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human PKN1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5585
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.