Learn More
Description
Specifications
Specifications
| Antigen | PKN1 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Formulation | Whole antisera with 0.05% Sodium Azide |
| Gene Accession No. | Q8TCW9 |
| Gene Alias | DBK, EC 2.7.11, MGC46204, PAK1Serine-threonine protein kinase N, PKNPKN-ALPHA, PRK1PAK-1, PRKCL1protein kinase C-related kinase 1, Protease-activated kinase 1, Protein kinase C-like 1EC 2.7.11.13, Protein kinase C-like PKN, protein kinase N1, Protein kinase PKN-alpha, Protein-kinase C-related kinase 1, serine/threonine protein kinase N, serine/threonine-protein kinase N1, serine-threonine kinase N |
| Gene Symbols | PKN1 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
