Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN2 Antibody (20A8), Novus Biologicals™
SDP

Catalog No. NB298218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB298218 50 μL
NB298219 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB298218 Supplier Novus Biologicals Supplier No. NBP32637950UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

PKN2 Monoclonal antibody specifically detects PKN2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry

Specifications

Antigen PKN2
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone 20A8
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias cardiolipin-activated protein kinase Pak2, EC 2.7.11, EC 2.7.11.13, Pak-2, PAK2, PRK2MGC150606, PRKCL2PRO2042, Protein kinase C-like 2MGC71074, protein kinase N2, Protein-kinase C-related kinase 2, serine/threonine-protein kinase N2
Host Species Rabbit
Immunogen A synthesized peptide derived from Human PKN2 [UniProt Q16513]
Purification Method Affinity purified
Quantity 50 μL
Regulatory Status RUO
Research Discipline Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 5586
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.