Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKN3 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP130102R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP130102R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP130102R Supplier Novus Biologicals Supplier No. NBP130102R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKN3 Polyclonal specifically detects PKN3 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.

Specifications

Antigen PKN3
Applications Western Blot, Immunoassay
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.11, EC 2.7.11.13, PKNbeta, protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, RP11-545E17.1, serine/threonine-protein kinase N3
Gene Symbols PKN3
Host Species Rabbit
Immunogen maps to a region between residue 225 and 275 of human protein kinase N3 using the numbering given in entry NP_037487.2
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 29941
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.