Learn More
Description
Specifications
Specifications
| Antigen | PKN3 |
| Applications | Western Blot, Immunoassay |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Dilution | Western Blot, Knockout Validated |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | EC 2.7.11, EC 2.7.11.13, PKNbeta, protein kinase N3, Protein kinase PKN-beta, Protein-kinase C-related kinase 3, RP11-545E17.1, serine/threonine-protein kinase N3 |
| Gene Symbols | PKN3 |
| Host Species | Rabbit |
| Immunogen | maps to a region between residue 225 and 275 of human protein kinase N3 using the numbering given in entry NP_037487.2 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
