Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PKR, p Thr446 Antibody, Novus Biologicals™
SDP

Catalog No. NB017877 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017877 0.025 mL
NB100821561 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017877 Supplier Novus Biologicals Supplier No. NB100821560.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PKR (p Thr446) Polyclonal specifically detects PKR (p Thr446) in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen PKR (p Thr446)
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS [pH 7.4] 150mM NaCl, and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide
Gene Accession No. P19525
Gene Alias double stranded RNA activated protein kinase, EC 2.7.11.1, eIF-2A protein kinase 2, EIF2AK1, eukaryotic translation initiation factor 2-alpha kinase 2P1/eIF-2A protein kinase, interferon-induced, double-stranded RNA-activated protein kinase, interferon-inducible double stranded RNA dependent, interferon-inducible elF2alpha kinase, Interferon-inducible RNA-dependent protein kinase, PKRp68 kinase, PRKRMGC126524, Protein kinase RNA-activated
Gene Symbols EIF2AK2
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human PKR around the phosphorylation site of threonine 446 (K-R-TP-R-S).
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Apoptosis, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5610
Test Specificity PKR (specific to Phospho-Thr446) detects endogenous levels of PKR only when phosphorylated at threonine 446.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.