Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5139916
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA5139916 100 μL
1 options
This item is not returnable. View return policy

Catalog No. PIPA5139916

Supplier: Invitrogen™ PA5139916

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: PVHSTMPGTV TSSCSPGNNS RPSSPGSGLH ASSPTASQNN SKAAVNSASS FNSSGSWYRW NHSTYL Highest antigen sequence indentity to the following orthologs: Mouse - 93%, Rat - 90%.

This gene encodes a member of the POU domain family of transcription factors. POU domain transcription factors bind to a specific octamer DNA motif and regulate cell type-specific differentiation pathways. The encoded protein is primarily expressed in the epidermis, and plays a critical role in keratinocyte proliferation and differentiation. The encoded protein is also a candidate tumor suppressor protein, and aberrant promoter methylation of this gene may play a role in cervical cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Sep 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU2F3
Gene Accession No. Q9UKI9
Gene Alias 11-Oct; Epoc-1; FLJ40063; LOC553427 protein; MGC126698; Oct11; oct-11; Oct-11a; octamer-binding protein 11; Octamer-binding transcription factor 11; OTF11; OTF-11; PLA1; PLA-1; POU class 2 homeobox 3; POU domain transcription factor OCT11a; POU domain, class 2, transcription factor 3; POU transcription factor; POU2F3; Rov 1 pou-homeodomain protein; Skin; Skin-1a; Skn-1a; Skn-li; Transcription factor PLA-1; transcription factor Skn-1; zgc:136377; zgc:158534
Gene Symbols POU2F3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human POU2F3. Recombinant protein control fragment (Product #RP-109346).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.