Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G12B Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB325624 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB325624 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB325624 Supplier Novus Biologicals Supplier No. NBP299414MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLA2G12B Polyclonal antibody specifically detects PLA2G12B in Mouse samples. It is validated for Western Blot

Specifications

Antigen PLA2G12B
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias group XIIB secretory phospholipase A2-like protein, group XIII secreted phospholipase A2, Group XIII secretory phospholipase A2-like protein, GXIIBMGC138151, GXIII sPLA2-like, GXIIIsPLA2, phospholipase A2, group XIIB, phospholipase A2, group XIII, PLA2G13, sPLA2-GXIIB
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse PLA2G12B.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84647
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.