Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G12B Antibody, Novus Biologicals™
SDP

Catalog No. NBP231685 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP231685 0.1 mL
NB397590 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP231685 Supplier Novus Biologicals Supplier No. NBP231685
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLA2G12B Polyclonal specifically detects PLA2G12B in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PLA2G12B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9BX93
Gene Alias group XIIB secretory phospholipase A2-like protein, group XIII secreted phospholipase A2, Group XIII secretory phospholipase A2-like protein, GXIIBMGC138151, GXIII sPLA2-like, GXIIIsPLA2, phospholipase A2, group XIIB, phospholipase A2, group XIII, PLA2G13, sPLA2-GXIIB
Gene Symbols PLA2G12B
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: DTEESYSDWGLRHLRGSFESVNSYFDSFLELLGGKNGVCQYRCRYGKAPMPRPGYKPQEPNGCGSYFLGLKVPESM
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 84647
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.