Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G16/HRASLS3 Antibody (OTI1A5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301945 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301945 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301945 Supplier Novus Biologicals Supplier No. NBP273454APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLA2G16/HRASLS3 Monoclonal antibody specifically detects PLA2G16/HRASLS3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen PLA2G16/HRASLS3
Applications Western Blot, Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
Clone OTI1A5
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias Adipose-specific phospholipase A2, AdPLAHRASLS3, EC 3.1.1.4, group XVI phospholipase A2, HRAS-like suppressor 3Ca-independent phospholipase A1/2, H-REV107-1, HREV107-3adipose-specific PLA2, HREV107H-rev 107 protein homolog, MGC118754, phospholipase A2, group XVI, Renal carcinoma antigen NY-REN-65
Host Species Mouse
Immunogen Full length human recombinant protein of human PLA2G16(NP_009000) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 11145
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.