Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2G4A Antibody, Novus Biologicals™
SDP

Catalog No. NBP152050 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP152050 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP152050 Supplier Novus Biologicals Supplier No. NBP152050
Add to Cart
Add to Cart

Goat Polyclonal Antibody

PLA2G4A Polyclonal specifically detects PLA2G4A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen PLA2G4A
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/mL, Flow Cytometry 10 ug/mL, Immunohistochemistry 3.75 ug/mL, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 3.75 ug/mL, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_077734.1
Gene Alias calcium-dependent phospholipid-binding protein, cPLA2, cPLA2-alpha, lysophospholipase, MGC126350, phosphatidylcholine 2-acylhydrolase, Phospholipase A2 group IVA, phospholipase A2, group IVA (cytosolic, calcium-dependent), PLA2G4cytosolic phospholipase A2
Gene Symbols PLA2G4A
Host Species Goat
Immunogen Peptide with sequence C-QEKTFRQQRKEHIR corresponding to internal region according to NP_077734.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5321
Test Specificity Cross reacts with Human and Mouse. Expected from sequence similarity: Rat.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.