Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLA2R1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146116 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146116 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146116 Supplier Novus Biologicals Supplier No. NBP146116
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

PLA2R1 Polyclonal specifically detects PLA2R1 in Human samples. It is validated for Western Blot, Flow Cytometry, Peptide ELISA.

Specifications

Antigen PLA2R1
Applications Flow Cytometry, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, Peptide ELISA 1:1000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q13018
Gene Alias 180 kDa secretory phospholipase A2 receptor, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180kD, secretory phospholipase A2 receptor
Gene Symbols PLA2R1
Host Species Goat
Immunogen Peptide with sequence C-EKKKDTPKQHGT corresponding to internal region according to NP_031392.3, NP_001007268.1, NP_001182570.1.
Molecular Weight of Antigen 153 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22925
Test Specificity This antibody is expected to recognize all reported isoforms (NP_031392.3; NP_001007268.1; NP_001182570.1).
Target Species Human, Pig
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.