Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLAC4 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB435981 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB435981 0.025 mg
NBP241125 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB435981 Supplier Novus Biologicals Supplier No. NBP2411250.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLAC4 Polyclonal specifically detects PLAC4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen PLAC4
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, ELISA, Immunocytochemistry/Immunofluorescence 10 -20 μg/mL
Formulation PBS with 0.02% Sodium Azide
Gene Alias C21orf115, Chromosome 21 Open Reading Frame 115, D21S418E, Placenta-Specific 4, Placenta-Specific Protein 4, PRED78
Gene Symbols PLAC4
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human PLAC4. The immunogen is located within amino acids 20 - 70 of PLAC4. Amino Acid Squence: DTLHPRHRHSSPWTPH
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 191585
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.