Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Lactogen/CSH1 Antibody (OTI2G4), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB301934 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301934 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301934 Supplier Novus Biologicals Supplier No. NBP273457APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Placental Lactogen/CSH1 Monoclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Placental Lactogen/CSH1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2G4
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin
Host Species Mouse
Immunogen Full length human recombinant protein of human CSH1(NP_072166) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1442
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.