Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Placental Lactogen/CSH1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB323485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323485 Supplier Novus Biologicals Supplier No. NBP299840PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Placental Lactogen/CSH1 Polyclonal antibody specifically detects Placental Lactogen/CSH1 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen Placental Lactogen/CSH1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Choriomammotropin, chorionic somatomammotropin hormone, chorionic somatomammotropin hormone 1 (placental lactogen), CS-1, CSAchorionic somatomammotropin A, CSH2, CSMT, FLJ75407, hCS-A, Lactogen, Placental lactogen, PLchoriomammotropin
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human Placental Lactogen/CSH1 (Accession#: NP_001308.1; Met1-Phe217)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cytokine Research, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1442
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.