Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Purified human placental lactogen
Molecular Weight: 21kDa
Positive Control: hPL recombinant protein, Placenta
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Specifications
| Antigen | Placental Lactogen (hPL) |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Rabbit |
| Immunogen | Purified human placental lactogen |
| Quantity | 1 mL |
| Regulatory Status | IVD |
| Research Discipline | Steroids Hormones and Growth Factors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.