Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Placental Lactogen (hPL) Rabbit Polyclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ Placental Lactogen (hPL), Rabbit Polyclonal Antibody.
Specifications
Antigen | Placental Lactogen (hPL) |
---|---|
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB9067P0
|
Epredia
RB9067P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
RB9067P
|
Epredia
RB9067P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB9067R1
|
Epredia
RB9067R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB9067R7
|
Epredia
RB9067R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
Human placental lactogen (hPL) is known to originate in the syncytiotrophoblasts. Ultrastructural localization indicates that placental lactogen is stored in small morphologically distinct cytoplasmic granules within these cells and has a different secretory mechanism from other hormonal peptides such as human chorionic gonadotropin. The level of expression of placental lactogen through pregnancy increases gradually to term.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested.
Epitope: Not known
Immunogen: Purified human placental lactogen
Molecular Weight: 21kDa
Positive Control: hPL recombinant protein, Placenta
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
Placental Lactogen (hPL) | |
Polyclonal | |
Rabbit | |
Steroids Hormones and Growth Factors | |
Purified human placental lactogen | |
Primary |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
IVD | |
Human | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title