Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLAGL1 Antibody (8F9D12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237344
Description
PLAGL1 Monoclonal specifically detects PLAGL1 in Human samples. It is validated for Western Blot, ELISA.Specifications
PLAGL1 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
DKFZp781P1017, LOT-1, LOT1ZACLost on transformation 1, MGC126275, MGC126276, PLAG-like 1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, ZAC tumor supressor, ZAC1, zinc finger protein PLAGL1 | |
Mouse | |
50.8 kDa | |
0.1 mL | |
Apoptosis, Neuronal Stem Cell Markers, Stem Cell Markers | |
5325 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
8F9D12 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
PLAGL1 | |
Purified recombinant fragment of human PLAGL1 (AA: 118-222) expressed in E. Coli. | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction