Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLAGL1 Antibody (8F9D12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | PLAGL1 |
---|---|
Clone | 8F9D12 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000 |
Applications | Western Blot, ELISA |
Description
PLAGL1 Monoclonal specifically detects PLAGL1 in Human samples. It is validated for Western Blot, ELISA.Specifications
PLAGL1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
RUO | |
Human | |
DKFZp781P1017, LOT-1, LOT1ZACLost on transformation 1, MGC126275, MGC126276, PLAG-like 1, pleiomorphic adenoma gene-like 1, pleiomorphic adenoma gene-like protein 1, Pleiomorphic adenoma-like protein 1, Tumor supressor ZAC, ZAC tumor supressor, ZAC1, zinc finger protein PLAGL1 | |
PLAGL1 | |
IgG2b | |
Affinity Purified |
8F9D12 | |
Western Blot 1:500 - 1:2000, ELISA 1:10000 | |
Monoclonal | |
Mouse | |
Apoptosis, Neuronal Stem Cell Markers, Stem Cell Markers | |
PBS with 0.05% Sodium Azide | |
5325 | |
Purified recombinant fragment of human PLAGL1 (AA: 118-222) expressed in E. Coli. | |
Primary | |
50.8 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title