Learn More
Kemtec™ Plant Cancer Kit
SCI_ED
Catalog No. S27529 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
9 to 14
Catalog No. Grade
S27529 9 to 14
1 options
Catalog No. S27529 Supplier Kemtec™ Supplier No. 2230
Add to Cart
Add to Cart
Kemtech™ Plant Cancer Kit allows students to investigate cancer in the laboratory.
  • Students safely explore aspects of cancerous cell growth with this self-contained kit.
  • A fresh carrot is required but not provided.
  • Experiments enable students to visualize the differences between normal cells and cancerous cells, describe similarities and differences between animal and plant cancers, identify cancer cells, identify a plant cancer or gall, and research current theories on the causes of cancer.
  • Students grow their own sunflower plants; then induce crown gall by inoculating plant tissue with Agrobacterium tumefaciens (reply card item ships upon request and only in Continental US) which is required for inducing the cancerous growth.
  • Students then do a Koch’s Postulates Experiment: Inducing Cancer in Carrot Tissue.
  • Allow 3 to 4 weeks if growing plants prior to innoculation and 3 to 4 weeks for tumor growth.
  • Experiment requires approximately one hour of lab time.
  • Designed for 24 students.
  • Recommended in conjunction with Koch's Postulate Experiment so as to use the same Agrobacterium tumefaciens culture or sub-culturing for later use.
  • Kit requires a free USDA permit for handling Agrobacterium tumefaciens to be obtained before the organism can be shipped.
  • Meets National Standards for Grade Levels 9 to 14
Specifications
Grade 9 to 14
Age 14 to 20
Includes Instructor's manual, Dry nutrient agar, Petri dishes, Spray bottles, Agar packet, Dissecting needles, Innoculating loops, Plant cancer flash drive, Sunflower seeds, Prepaid Agrobacterium Living Coupon
Weight (English) 3 lb.
Dimensions (L x W x H) 18 x 12 x 6 in.