Learn More
Plasma Cell Antibody (LIV3G11 (7B18)), Alexa Fluor™ 647, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Plasma Cell |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | LIV3G11 (7B18) |
| Conjugate | Alexa Fluor 647 |
| Dilution | Immunohistochemistry-Paraffin |
| Gene Alias | Plasma |
| Host Species | Mouse |
| Immunogen | Pancreatic cancer related serum mucin was used as the immunogen for the Plasma Cell antibody. |
| Purification Method | Protein A or G purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.