Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasma Cell Antibody (LIV3G11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP25025100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP25025100 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP25025100 Supplier Novus Biologicals Supplier No. NBP250251AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plasma Cell Monoclonal specifically detects Plasma Cell in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen Plasma Cell
Applications Immunohistochemistry
Classification Monoclonal
Clone LIV3G11
Conjugate Alexa Fluor 700
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Plasma
Host Species Mouse
Immunogen Pancreatic cancer related serum mucin
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Test Specificity Does not react with normal B cells. Reacts with normal and malignant human plasma cells
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.