Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasminogen Antibody (SBF1 C1.21), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB1002791FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1002791FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1002791FR Supplier Novus Biologicals Supplier No. NB1002791FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Plasminogen Monoclonal specifically detects Plasminogen in Human samples. It is validated for ELISA.

Specifications

Antigen Plasminogen
Applications ELISA
Classification Monoclonal
Clone SBF1 C1.21
Conjugate DyLight 680
Dilution ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Gene Symbols PLG
Host Species Mouse
Immunogen Human Plasminogen
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5340
Test Specificity Does not cross react with HGF1/MSP or HGF/SF. Cross reacts with plasmin
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.