Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plasmolipin Antibody, Novus Biologicals™
SDP

Catalog No. NB397727 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397727 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397727 Supplier Novus Biologicals Supplier No. NBP2224650.025MG

Rabbit Polyclonal Antibody

Plasmolipin Polyclonal specifically detects Plasmolipin in Rat samples. It is validated for Western Blot.

Specifications

Antigen Plasmolipin
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation PBS and 1 mg/mL BSA with 0.05% Sodium Azide
Gene Alias Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin)
Gene Symbols PLLP
Host Species Rabbit
Immunogen Synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51090
Target Species Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.