Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Plasmolipin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$173.10
Specifications
Antigen | Plasmolipin |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 2 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Plasmolipin Polyclonal specifically detects Plasmolipin in Rat samples. It is validated for Western Blot.Specifications
Plasmolipin | |
Western Blot 2 μg/mL | |
Polyclonal | |
Rabbit | |
Rat | |
Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin) | |
PLLP | |
IgG | |
Affinity Purified |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
PBS and 1 mg/mL BSA with 0.05% Sodium Azide | |
51090 | |
Synthetic peptides corresponding to residues C D(127) L T S L R G (S/T) R P Y N Q R S(141) of human and rat Plasmolipin. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title