Learn More
American Educational Products Plastic Column Kit
Shop All American Educational Products LLC Products
Description
Includes
tube, drainage fixture, screen, and teacher's guide.
Learn how the movement of water is affected by substrate!
How do porosity and permeability affect the distribution of aquifers? Help students study these important concepts and integrate math measurement and percentage skills. Bead set is optional. Grades 6-12.
Specifications
Specifications
| Grade | 6 to 12 |
| Product Type | 7 mm Beads |