Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plastin L Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB101001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB101001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB101001 Supplier Novus Biologicals Supplier No. NBP297134C

Rabbit Polyclonal Antibody

Plastin L Polyclonal specifically detects Plastin L in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Plastin L
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias CP64, DKFZp781A23186, FLJ25423, FLJ26114, FLJ39956, LC64PPLS2bA139H14.1 (lymphocyte cytosolic protein 1 (L-plastin)), LCP-1, LPL, L-plastin, L-plastin (Lymphocyte cytosolic protein 1) (LCP-1) (LC64P), Lymphocyte cytosolic protein 1, lymphocyte cytosolic protein 1 (L-plastin), Lymphocyte cytosolic protein-1 (plasmin), plastin 2, plastin-2
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Plastin L fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3936
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.