Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Platelet Monoclonal Antibody (AIP21), FITC
Rat Monoclonal Antibody
$247.50 - $483.50
Specifications
Antigen | Platelet |
---|---|
Clone | AIP21 |
Concentration | 0.1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA517900
|
Invitrogen™
MA517900 |
100 μg |
Each for $247.50
|
|
|||||
PIMA517901
|
Invitrogen™
MA517901 |
300 μg |
Each for $483.50
|
|
|||||
Description
Platelet Monoclonal Antibody for Flow
Platelets are nonproliferative and terminally differentiated cells. Platelets offer an attractive model system to study the various biochemical events leading to structural and functional alterations in activated cells. When platelets are exposed to stimuli, they are activated, undergo a dramatic shape change, adhere to each other, and aggregate.Specifications
Platelet | |
0.1 mg/mL | |
Flow Cytometry | |
FITC | |
Rat | |
RUO | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
Mouse Platelets. | |
Antibody |
AIP21 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
Monoclonal | |
Liquid | |
IgM | |
Mouse | |
Thrombocytes | |
Primary | |
Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title