Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Platinum(II) chloride, 73% Pt
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 581.0°C |
| Color | Brown to Gray-Green |
| Physical Form | Powder |
| Quantity | 250 mg |
| Linear Formula | PtCl2 |
| Fieser | 02,272 |
| Merck Index | 15,7643 |
| Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid,insoluble in alcohol and ether |
| Formula Weight | 266 |
| Percent Purity | 73% (Pt) |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. |
| Precautionary Statement | GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective |
| EINECSNumber | 233-034-1 |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction