Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLBD2 Rabbit anti-Human, DyLight 650, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB086899 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086899 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086899 Supplier Novus Biologicals Supplier No. NBP300235C

Rabbit Polyclonal Antibody

PLBD2 Polyclonal specifically detects PLBD2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen PLBD2
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Formulation 50 mM Sodium Borate
Gene Alias LAMA-like protein 2,76 kDa protein, lamina ancestor homolog 2, mannose-6-phosphate protein associated protein p76, P76, phospholipase B domain containing 2, phospholipase B domain-containing protein 2, phospholipase B-like 2 32 kDa form, phospholipase B-like 2 45 kDa form, PLB homolog 2, putative phospholipase B-like 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant PLBD2 (Q8NHP8; Met1-Asp589).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 196463
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.