Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleckstrin-2 Antibody (OTI5D3), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273467A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273467A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273467A30 Supplier Novus Biologicals Supplier No. NBP273467AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pleckstrin-2 Monoclonal specifically detects Pleckstrin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Pleckstrin-2
Applications Western Blot
Classification Monoclonal
Clone OTI5D3
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2
Gene Symbols PLEK2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 132-353 of human Pleckstrin-2 (NP_057529) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26499.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.