Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleckstrin-2 Antibody (OTI5D3) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273467 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273467 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273467 Supplier Novus Biologicals Supplier No. NBP273467
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pleckstrin-2 Monoclonal specifically detects Pleckstrin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen Pleckstrin-2
Applications Western Blot
Classification Monoclonal
Clone OTI5D3
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2
Gene Symbols PLEK2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 132-353 of human PLEK2 (NP_057529) produced in E.coli.
Molecular Weight of Antigen 39.8 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 26499
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.