Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleckstrin Antibody (OTI6H8), PE/Cy7, Novus Biologicals™
SDP

Catalog No. N273464PEC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273464PEC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273464PEC7 Supplier Novus Biologicals Supplier No. NBP273464PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pleckstrin Monoclonal specifically detects Pleckstrin in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Pleckstrin
Applications Flow Cytometry
Classification Monoclonal
Clone OTI6H8
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ27168, p47, P47Platelet 47 kDa protein, pleckstrin
Gene Symbols PLEK
Host Species Mouse
Immunogen Full length recombinant protein of human Pleckstrin (NP_002655) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5341.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.