Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEK Mouse anti-Human, Porcine, Clone: 1D5C2, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | PLEK |
---|---|
Clone | 1D5C2 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6858
![]() ![]() |
Proteintech Group Inc
664311IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Major protein kinase C substrate of platelets.Specifications
PLEK | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P47, Platelet 47 kDa protein, pleckstrin, PLEK | |
PLEK | |
IgG1 | |
Protein G | |
Antibody |
1D5C2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Pig | |
P08567 | |
100516257, 5341 | |
PLEK Fusion Protein Ag3187 | |
Primary | |
-20°C | |
PLEK |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title