Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHA4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15768420 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15768420 20 μL
NBP157684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15768420 Supplier Novus Biologicals Supplier No. NBP15768420UL

Rabbit Polyclonal Antibody

PLEKHA4 Polyclonal specifically detects PLEKHA4 in Human samples. It is validated for Western Blot.

Specifications

Antigen PLEKHA4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q9H4M7
Gene Alias PEPP-1, PEPP1phosphoinositol 3-phosphate binding protein-1, PH domain-containing family A member 4, phosphoinositol 3-phosphate-binding PH domain protein 1, Phosphoinositol 3-phosphate-binding protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 4, pleckstrin homology domain-containing family A member 4, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 4
Gene Symbols PLEKHA4
Host Species Rabbit
Immunogen Synthetic peptides corresponding to PLEKHA4(pleckstrin homology domain containing, family A member 4) The peptide sequence was selected from the middle region of PLEKHA4. Peptide sequence TEPDSPSPVLQGEESSERESLPESLELSSPRSPETDWGRPPGGDKDLASP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57664
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.