Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | PLEKHA4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15768420
![]() |
Novus Biologicals
NBP15768420UL |
20 μL |
Each for $206.00
|
|
|||||
NBP157684
![]() |
Novus Biologicals
NBP157684 |
100 μL |
Each for $487.50
|
|
|||||
Description
PLEKHA4 Polyclonal specifically detects PLEKHA4 in Human samples. It is validated for Western Blot.Specifications
PLEKHA4 | |
Polyclonal | |
Rabbit | |
Q9H4M7 | |
57664 | |
Synthetic peptides corresponding to PLEKHA4(pleckstrin homology domain containing, family A member 4) The peptide sequence was selected from the middle region of PLEKHA4. Peptide sequence TEPDSPSPVLQGEESSERESLPESLELSSPRSPETDWGRPPGGDKDLASP. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
PEPP-1, PEPP1phosphoinositol 3-phosphate binding protein-1, PH domain-containing family A member 4, phosphoinositol 3-phosphate-binding PH domain protein 1, Phosphoinositol 3-phosphate-binding protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 4, pleckstrin homology domain-containing family A member 4, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 4 | |
PLEKHA4 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title