Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHA7 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB336846 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB336846 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB336846 Supplier Novus Biologicals Supplier No. NBP276328APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLEKHA7 Polyclonal antibody specifically detects PLEKHA7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen PLEKHA7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZp686M22243, PH domain-containing family A member 7, pleckstrin homology domain containing, family A member 2, pleckstrin homology domain containing, family A member 7, pleckstrin homology domain-containing family A member 7
Host Species Rabbit
Immunogen Partial recombinant human PLEKHA7 protein (amino acids 821-1121) [UniProt Q6IQ23}
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144100
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.