Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEKHA7 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB009373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009373 Supplier Novus Biologicals Supplier No. NBP276328B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLEKHA7 Polyclonal specifically detects PLEKHA7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen PLEKHA7
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp686M22243, PH domain-containing family A member 7, pleckstrin homology domain containing, family A member 2, pleckstrin homology domain containing, family A member 7, pleckstrin homology domain-containing family A member 7
Gene Symbols PLEKHA7
Host Species Rabbit
Immunogen Partial recombinant human PLEKHA7 protein (amino acids 821-1121) [UniProt Q6IQ23}
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.